Would you like to log in again?
VWR® Adjustable Spray Bottle
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
23609-182
23609-182PK 82.63 CAD InStock
23609-182
VWR® Adjustable Spray Bottle
Bottles Spray Bottles
The bottle's adjustable sprayer permits a range of positioning from fine mist to large stream delivery to accommodate a variety of washing applications.
  • HDPE and polypropylene construction
  • Chemical resistant and unbreakable
  • Instant-off release eliminates messy dripping
The chemical-resistant and unbreakable bottle is constructed of HDPE and the sprayer is made from polypropylene. The pump trigger is comfortable for one-handed operation and features an instant-off release that eliminates messy dripping. Closed position on sprayer maintains solutionMore Product Information
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms