Would you like to log in again?
MICROLITER® 700 Series Syringes, Hamilton
Supplier: Hamilton
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
80301 80401 80501 80601 80701 80801 80400 80500 80600 80700 80800 80865 80765 87900 87930 80330 80336 80430 80530 80630 80730 80830 80365 80465 80565 80665 80621 80419 80521 80339 80739 80439 80539 80639 80839 80366 80300 80342 80408 80508 80608 80708 80808 87919 80383 87908 80308
60371-000EA 158.93 CAD InStock
60371-000 60371-011 60371-022 60371-044 60371-066 60371-088 60372-105 60373-028 60373-040 60373-061 60373-083 60373-323 60373-345 60373-370 60373-375 60373-470 60373-471 60373-505 60373-538 60373-560 60373-593 60373-629 60373-946 60373-957 60373-968 60373-979 60373-980 60376-170 60378-467 72310-434 72310-442 72310-444 74980-002 74980-004 74980-006 60372-025 60372-091 10118-396 10192-052 10192-054 10192-056 10192-058 10192-072 10192-092 10192-094 10192-148 10192-150
MICROLITER® 700 Series Syringes, Hamilton
Syringes Syringes for Chromatography
The 700 series syringe is the original hand-fitted Hamilton syringe
Cemented needles (N) for low volume syringes feature the needles cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable. Syringe 60373-775 features a special cemented needle (SN) with a termination that is the same as a cemented needle termination, except it allows for a variety of user-defined gauges,More Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms