Kilo Scale Bubbler, Ace Glass
Supplier: Ace Glass
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7420-01
76214-458EA 2516.07 CAD
76214-458
Kilo Scale Bubbler, Ace Glass
Bubblers
Kilo scale bubbler for inerting or purging large reaction vessels.
  • High-quality borosilicate glass
  • Ace-Safe™ hose connections eliminate the safety hazard of broken glass hose connections
Bubbler features a ground glass check valve which prevents oil or air from being pulled into the system.
Ace-Safe™ hose connections for 3/8" ID tubing.