Syringes for Dosage of Gas and Liquids, H
Supplier: INNOVATIVE LABOR SYSTEME GMBH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2600501
77506-134EA 84.93 CAD
77506-134
Syringes for Dosage of Gas and Liquids, H
Syringes Syringes for Chromatography
Sample dosage for Gas Chromatography (GC).
  • HPLC valves
  • Autosamplers
  • Dosage of aggressive fluids
  • Glass barrel: Borosilicate 3.3
Plunger: Stainless steel with PTFE-seal
Precision: <±1% of the volume