Sharps Containers, Stackable, Medegen
Supplier: Medegen Medical Products, LLC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8710
89166-424CS 449.62 CAD
89166-424
Sharps Containers, Stackable, Medegen
Bins Sharps Containers
Large multi-purpose sharps containers are suitable for OR and dialysis units, and contain locks for final disposal.
Containers feature a tortuous path lid with large and small openings for infectious waste of various types.