Would you like to log in again?
VWR® Plastic Laminate Worksurface
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AMCLWB3130H AMCLWB3130M AMCLWB3136H AMCLWB3136M AMCLWB3148H AMCLWB3148M AMCLWB3160H AMCLWB3160M AMCLWB3172H AMCLWB3172M AMCLWB3530H AMCLWB3530M AMCLWB3536H AMCLWB3536M AMCLWB3548H AMCLWB3548M AMCLWB3560H AMCLWB3560M AMCLWB3572H AMCLWB3572M
AMCLWB3130HEA 883.57 CAD InStock
AMCLWB3130H AMCLWB3130M AMCLWB3136H AMCLWB3136M AMCLWB3148H AMCLWB3148M AMCLWB3160H AMCLWB3160M AMCLWB3172H AMCLWB3172M AMCLWB3530H AMCLWB3530M AMCLWB3536H AMCLWB3536M AMCLWB3548H AMCLWB3548M AMCLWB3560H AMCLWB3560M AMCLWB3572H AMCLWB3572M
VWR® Plastic Laminate Worksurface
Furniture Tables and Workbenches
Laminate worksurfaces are composed of high-pressure plastic laminate.
  • For High-Column worksurface two or three cut-outs accomodate 76" high columns, depending on bench width
  • High-Column worksurfaces may be used with sitting/bench height carts when height-adjustment is required
Each worksurface has three square edges and one full 180-degree wrapped front edge.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms