Would you like to log in again?
VWR® Petri Dishes, Nesting Alpine
Supplier: VWR International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CA73370-042
CA73370-042CS 309.11 CAD InStock
CA73370-042
VWR® Petri Dishes, Nesting Alpine
Culture Dishes Petri Dishes
The unique recessed lid on these dishes offers excellent stacking stability and saves space.
  • Recessed lid design
  • Excellent stackability
  • Compact design
  • Flat bottom
They work with most models of media pouring equipment
The flat bottom allows a reduction in the amount of media required. Patented cross ribs on the bottom make these dishes resilient to warpage when media is poured at a high temperature. These dishes are excellent for performing colony counts because the cross ribs also provide a useful border for making quadrents.
Available with fourMore Product Information
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms