Would you like to log in again?
Masterflex® L/S® MasterSense™ Pump Systems
Supplier: MASTERFLEX LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
77920-42 77920-42 77920-42 77920-42 77920-42 77920-42 77920-42 77920-42 77920-42 77920-40 77976-00 77976-02 77976-10 77976-12 77920-50 77920-52 77920-50 77976-02-AU 77976-00-AU 77976-12-AU 77976-10-AU 77976-00-DK 77976-02-DK 77976-10-DK 77976-12-DK 77920-50 77976-00-IN 77976-02-IN 77976-10-IN 77976-12-IN 77920-50 77976-00-IL 77976-02-IL 77976-10-IL 77976-12-IL 77920-50 77976-00-EU 77976-02-EU 77976-10-EU 77976-12-EU 77920-50 77976-00-UK 77976-02-UK 77976-10-UK 77976-12-UK 77920-50 77976-00-CH 77976-02-CH 77976-10-CH 77976-12-CH 77920-50 77976-00-IT 77976-02-IT 77976-10-IT 77976-12-IT 77920-50
MFLX77920-42EA 9085 CAD InStock
MFLX77920-42 MFLX77920-42-AU MFLX77920-42-DK MFLX77920-42-IN MFLX77920-42-IL MFLX77920-42-EU MFLX77920-42-UK MFLX77920-42-CH MFLX77920-42-IT MFLX77920-40 MFLX77976-00 MFLX77976-02 MFLX77976-10 MFLX77976-12 MFLX77920-50 MFLX77920-52 MFLX77920-50-AU MFLX77976-02-AU MFLX77976-00-AU MFLX77976-12-AU MFLX77976-10-AU MFLX77976-00-DK MFLX77976-02-DK MFLX77976-10-DK MFLX77976-12-DK MFLX77920-50-DK MFLX77976-00-IN MFLX77976-02-IN MFLX77976-10-IN MFLX77976-12-IN MFLX77920-50-IN MFLX77976-00-IL MFLX77976-02-IL MFLX77976-10-IL MFLX77976-12-IL MFLX77920-50-IL MFLX77976-00-EU MFLX77976-02-EU MFLX77976-10-EU MFLX77976-12-EU MFLX77920-50-EU MFLX77976-00-UK MFLX77976-02-UK MFLX77976-10-UK MFLX77976-12-UK MFLX77920-50-UK MFLX77976-00-CH MFLX77976-02-CH MFLX77976-10-CH MFLX77976-12-CH MFLX77920-50-CH MFLX77976-00-IT MFLX77976-02-IT MFLX77976-10-IT MFLX77976-12-IT MFLX77920-50-IT
Masterflex® L/S® MasterSense™ Pump Systems
Pumps Peristaltic Pumps
Complete pump systems for accurate fluid delivery with an intuitive touch-screen interface.
  • Precise, repeatable performance that is quick and easy to set up and program
  • 5'' (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Options for connectivity and monitoring include MasterflexLive, EtherNet/IP™, Profibus, or analog signal
  • Three-
More Product Information
Order Now
ORDER
RELATED INFORMATION
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms