Standard single-use products
Through VWR, part of Avantor, choose from over 100 standard products that have been designed to solve many common bioprocess applications. All products featured below are available for prompt delivery.
Not seeing what you're looking for? Our team of experts can apply their fluid transfer knowledge to design custom single-use solutions to meet your specific applications. Learn about our custom single-use solutions.
Product Selection
Masterflex® Single-Use Components
Tubing, fittings, clamps, sensors and more for single-use applications such as bioprocessing and beyond.
Avantor TopMixer 200 & TopMixer 500
Modular mixing systems for our biopharma processes with integrated formulation programming
Avantor Single-Use High Pressure Transfer Hose with SterilEnz™ Connections, Avantor Fluid Handling
Avantor's high-pressure single-use transfer hose with Avantor SterilEnz™ connections maintain the durability of high-pressure hose products while providing the benefits and flexibility of single-use assemblies.
Avantor Single-Use Bioprocessing Bags
Avantor single-use bioprocessing bags are designed to transfer and store your products throughout the drug-manufacturing process.
OmniTop™ single-use sampling and small volume transfer tubes
Pre-sterilized single-use tubes for aseptic sample transfer of biopharmaceutical products and reagents in a closed system.
WFI samplers with pre-gasketed fittings
Pre-attached SterilEnz®-II/AT gasket fittings provided added convenience when gathering aseptic samples from a WFI water drop.
Sterile single-use PETG bottle assemblies
Pre-sterilized for convenience and offered in a range of sizes with tubing that is completely weldeable/sealable.
Sterile single-use polycarbonate bottle assemblies
Designed to withstand temperatures from -20 to 65C (-4 to 149F) without losing integrity and offered in a range of convenient sizes.
Medical-grade single-use tank liners
Featuring an open top designed to fit in cylindrical vessels to avoid cleaning after media and buffer preparation.
SterilEnz®-II/AT pre-gasketed single-use fittings
Pre-gasketed design helps prevent contamination risks during assembly.
SterilEnz®-II/EC pre-gasketed polypropylene end caps
The only single-use connectors that come with a medical-grade silicone gasket mechanically attached to the fitting face.
Wire-reinforced silicone hose and labels
Ideal for biopharmaceutical and biomedical applications, with excellent bend radius and kink resistance.
Stainless steel wall-mounted hose racks
Conveniently store hose and components while enhancing safety by eliminating clutter in the work environment.
Looking for a custom solution?
Learn how we can help tailor a solution to meet the needs of your specific application. Contact us today.