Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, October 23. 
Cannabis and Hemp
In the rapidly growing Cannabis and Hemp industry, Avantor is your source to ensure a dependable supply chain of products from cultivation through packaging, providing solutions for growing, testing, extraction and formulation. Whether you are a Cannabis and Hemp testing laboratory, producer, or processor, Avantor's solutions have you covered.
Cannabis and Hemp Workflow
Featured products
From Plant to Patient
DWK Life Science is here to support the rapidly expanding Cannabis Market. Our comprehensive range of labware is your one source for testing, production and packaging.
Leucosep™ Tubes for Extraction
Pre-fritted 50mL tubes useful in separation VWR Catalog Number 89048-936 available in packs of 25 or cases of 300
MA160 Moisture Analyzer
Fully automated Infrared Moisture Analyzer for reliable and traceable cannabis moisture results during sample prep for analytics of THC content, CO2 extraction and microbial growth