Would you like to log in again?
Inlet Liners
Inlet Liners
Maintaining a clean injection port is critical to an inert flow path and reliable results. This involves the use of glass inlet liners, precision inlet seals, and reliable O-rings.
Agilent offers a wide variety of inlet liners for split splitless injections, cool on-column (COC) injections and multimode injections, with and without glass wool. If your samples require complete inertness, try any of the products coated with Agilent's exclusive Ultra Inert technology. We also provide a complete offering of liner O-rings and inlet liners for third-party instruments, making Agilent a one-stop shop for all your supplies.