Would you like to log in again?
Inlet Septa for Gas Chromatography
Inlet Septa for Gas Chromatography
Advanced Green inlet septa eliminate sticking and reduce coring. Bleed temperature optimized (BTO) inlet septa are preconditioned and ready to use with low-bleed, mass spec capillary columns. Our long-life septa are specially formed for extended use, meaning less downtime. Explore Agilent's complete portfolio of gas chromatography septa to ensure your GC is operating at peak performance.