Would you like to log in again?
PIG® HazMat Spill Kits - The only spill kits packed full of PIG HazMat Absorbents!
PIG® HazMat Spill Kits
WORLD-CLASS ABSORBENTS
Absorb the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide.
VARIETY OF CONTAINERS
Choices range from portable grab-and-go to mobile carts to overpack salvage drums.
PACKED FOR FAST RESPONSE
Kits are packed in the order contents are needed for fast response and access.
PIG® HazMat Spill Kit in High-Visibility Bag
Absorbs up to 9 gal., Absorbs Acids; Bases; Unknowns PIG Chemical Absorbents in an easy to see, fast to find, hi-viz bag.
PIG® HazMat Spill Kit in High-Visibility Caddy
Absorbs up to 14 gal., Absorbs Acids; Bases; Unknowns .Fully stocked, wheeled mobile cart features swing out front doors for easy access.
PIG® HazMat Spill Kit in 95-Gallon Wheeled Overpack Salvage Drum
Absorbs up to 63 gal., Absorbs Acids; Bases; Unknowns, UN1H2/X113/S Strong enough to earn a UN rating for shipping chemical spill cleanup waste.
PIG® HazMat Spill Kit in 20-Gallon Overpack Salvage Drum
Absorbs up to 11 gal., Absorbs Acids; Bases; Unknowns, UN1H2/X113/S Strong enough to earn a UN rating for shipping chemical spill cleanup waste.
PIG® Base Neutralizing Spill Kit in Bucket
Absorbs up to 4 gal., Absorbs Bases. Safely respond to corrosive spills with this specially equipped spill kit with neutralizing absorbents and PPE.
VWR Lab Spill Kit
Have your lab ready with this compact Kit. It’s packed with HAZ-MAT absorbents and PPE to safely contain and soak up small spills of chemicals.