Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, October 23. 
Puritan Medical Products
Puritan Medical Products
Puritan Medical Products Co., LLC, is over 100 years old and is a family-owned American company known worldwide as the trusted manufacturer of single-use products for controlled environments, diagnostics, specimen collection, environmental, forensics, genetics, medical, and microbiology media industries. Puritan is FDA-registered and ISO-certified. Puritan's products help customers collect, analyze, and transport specimens with the utmost confidence in the products’ integrity. All patented flock swabs and related products are manufactured in its Guilford, Maine and Pittsfield, Maine facilities. 
Puritan's patented HydraFlock® and PurFlock Ultra® flocked swabs have optimum absorption and elution capabilities and are the preferred swab for specimen collection and patient comfort. UniTranz-RT® (Universal transport media) is used to test for viruses, chlamydia, mycoplasma, and ureaplasma specimens at room temperature (RT) or long-term freezer storage. Puritan’s Opti-Swab® Modified Liquid Amies transport medium maintains viability of aerobic, anaerobic and fastidious bacteria. 
Puritan's media transport systems combined with high-performing swabs create the ideal product for specimen collection and transport. Puritan will always be in the forefront, meeting the needs of emerging technologies.