RheoCrimp
The RheoCrimp is the latest in easy to use cut and crimp separation technology. These crimps are very easy to use as they offer a wide opening range for capturing bulky liner material. The RheoCrimp Kit includes the tools needed for this crimp and cut processs, as well as a pack of 50 crimps. This starter kit comes in a high-quality carrying case and includes:
- Cable Tie Tensioner and Cutting Tool
- 50 High-Force Cable Ties
- Roll of GMP Tape
- Cutting Sheers for cutting between Cable-Ties
The RheoCrimp is an easy to use crimp with a wide, wrap-around design that achieves a tight cinch.
- 50 High-Force Cable Ties per package
- Wide-Range Crimp Capability
- Dual-Tie Design
- Easy Alignment Cut-Notch for Cutting Operation