Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Sunday, October 23. 
Ansell Chemical Resistant Gloves
Microflex 93-260
Introducing the Microflex 93-260, the new glove that's changing the face of chemical hand protection. With an innovative three layer design, Microflex 93-260 offers the reliable protection workers need when handling chemicals.
Innovative 3 Layer Design*
The glove’s unique, triple-layer construction features a soft interior specifically designed to enable easy donning and doffing. Its flexible middle layer delivers maximum protection against acids and bases, while the durable exterior delivers maximum protection against organic solvents. Because the glove’s tough exterior stands up longer to demanding physical and chemical conditions workers may change gloves less frequently resulting in enhanced productivity.
Tough Chemical Protection
The Microflex 93-260 provides such tough chemical protection that it has met requirements for both EN 374 certification in Europe, and KOSHA certification in Korea. When tested against aggressive chemicals like Sulfuric Acid, N Heptane, Hexane and Methanol, the Microflex 93-260 demonstrates impressive permeation breakthrough times – often outperforming competitors.
Unparalleled Comfort
Made with an innovative nitrile and neoprene composite material, the Microflex® 93-260 measures only 7.8 mil (0.19 mm) thick and delivers all the benefits of a single-use, splash resistant glove: exceptional comfort, fit, dexterity and grip. These attributes combined with Ergoform Design Technology help reduce hand and forearm muscle fatigue to support worker productivity, and reduce the likelihood of costly or dangerous accidents and spills.
* The process used to make this 3 layer glove is patent pending.
Flyers
Watch Our Videos
Introduction to
Microflex 93-260