Would you like to log in again?
Fixing and Holding Solutions
The preservation of all specimens involves the chemical alterations of the tissue proteins that inhibit bacterial and fungal growth that ultimately leads to decomposition. Fixation is a process in which a chemical fixative reduces tissues to an incorruptible state.
After fixation, specimens are rinsed in water and placed in a holding solution. Our preserving methods provide safe, worry-free handling of each specimen.
Holding Solutions
Ward’Safe
Once fixed, Ward's Science regular preserved materials are placed in Ward’Safe, a holding fluid that maintains the state of preservation.
  • Contains no formalin
  • Contains a formaldehyde scavenger that seeks out and binds to the free formaldehyde in formalin-containing fluids that may seep from specimens, effectively reducing the already minimal residual level.
  • Independent laboratory tests have determined Ward’Safe is non-sensitizing to skin. It is not a primary skin or eye irritant in accordance with Consumer Product Safety Commission.
  • All pail packed specimens are packed in Ward’Safe, with the exception of botanical specimens, which are packed in glycerin, and some arthropods, which are packed in alcohol.
100% Form-Free Specimens
Absolutely no formaldehyde is used during processing of our Form-Free specimens. This method involves a fixative which is totally consumed during the fixation process so that none of the chemical remains in the specimen. A 100% Form-Free specimen allows students and instructors that have concerns over formaldehyde exposure the opportunity to perform worry-free dissections.