Would you like to log in again?
Labware
Labware
When it comes to your laboratory needs, Sklar has you covered with a remarkable line of stainless steelware, plasticware, and brushes. Choose from a wide variety of products such as sterilization containers, instrument cassettes, instrument soaking trays, instrument protectors, medicine cups, sponge bowls, solution basins, hospital buckets, forcep jars, and more! All labware is produced to ensure superior performance and durability to meet the rigorous demands required by any healthcare and laboratory specialist.
To organize and secure the sterilization of your surgical instruments, Sklar has developed the The Sklarlite Sterilization Container System. The container system is available in various sizes and configurations to fit your needs and customizable through a range of colors, lids, and labels for simpler identification. Containers are stackable and lightweight for effortless handling, yet durable and reliable thanks to their anodized aluminum alloy construction. The system also utilizes silicone mats to minimize damage to delicate laparoscopic instruments and tamper-evident seals to prevent breaches in sterilization.