Would you like to log in again?
Spectrum offers BASF catalysts
BASF catalysts
BASF built on its world-renowned background in materials science and surface chemistry to create a range of efficient, high-performance precious metal powder catalysts. Its patented NanoSelect™ catalysts are defined by unimodal, nano-sized metal particles, increasing the available surface area of the metals to greatly boost catalytic activity. The innovative technology combines stabilizing and reducing functions to produce colloids that can be used with different support materials to create heterogeneous catalysts with unique catalytic behavior.