To secure a supply chain during a global constraint of silicone, it’s best to be flexible The global constraint of siloxane (a key ingredient in silicone) has caused extended lead times, increased pricing, and created headaches in supply chains across the market that are sourcing this material. Unfortunately, the constraint may continue for many months and with one major source of silicone tubing projecting another 12 months of restricted flow of siloxane. Over the past few decades, silicone’s dominance in the medical and pharmaceutical marketplace has expanded and various competitive elastic materials have been developed to grab some market share. The associated benefits are as numerous as the formulations available: reduced costs, longer pumping life (for those formulations designed to work in peristaltic pumps), expanded chemical and temperature compatibility, decreased gas permeability, and higher purity to name just a few. To navigate this sea of available silicone alternatives quickly, first determine the critical process parameters and the reason silicone was chosen, and then choose a source that can complete the fluid path for the need. The siloxane constraint in summary Naturally, the demand for silicone products will increase alongside the gains in the global economy, which have been robust. However, recently pharmaceutical, medical and construction industries have been expanding faster than the rate of the economy, leading to a spike in demand. This new demand is reflected in the facilities popping up in growth markets such as those in the Asia-Pacific region. In addition to the new demand increase, supply constraints have been introduced. Normally, if one region’s capacity was tight or experiencing issues with flow of components / ingredients needed for production, the solution was to explore other regions for excess capacity. However due to changes at facilities that produce siloxane (i.e., new environmental regulations, work stoppages, and a complete production facility being taken out of the global capacity) a ripple effect downstream can be observed. Raw material lead times and price have increased impacting the manufacturing plants that require the flow of siloxane to produce the products used in personal lives or to make other items for the global economy (i.e., vaccines, drugs, etc.). Choosing an alternative tubing formulation Unfortunately, in some cases the timing needed to source the required grade of silicone tubing does not match the production time line and an alternative formulation may be the necessary solution. To do this, one must first perform a review of the fluid path. Consider what is going on inside and outside the tubing—as there are two parts of chemical compatibility: what impact the fluid has on the materials around the fluid path, and the impact those materials have on the fluid being pumped. Other parameters to consider are the size to accommodate the needed flow rate, system and ambient pressure, temperature, accessibility, batch times, and necessary regulatory requirements. Some external parameters are welded joints and sterilization technique (if necessary). Internal considerations Primarily, chemical compatibility needs to be considered between the process media and the tube material. Make sure to consider every component of the fluid especially if it is in contact with the tubing for a long time. A quick search on the Internet for chemical compatibility charts of elastic materials will yield many results. Pick a few that fit the process and average their data. Most should agree with one another. If the fluid cannot be found or the process doesn’t fit the test parameters (most charts assume room temperature), there are other options. Some tubing suppliers offer a tubing chemical compatibility kit or samples of their formulations. This ensures the customer’s formulation of choice is compatible with the process. Masteflex has a tubing kit that recommends an immersion test where the sample tube’s dimensions and weight are recorded before immersion into the process media. The time for the sample immersion should be 48 hours to be sure of any potential long-term effects the process media have on the formulation. Once the period passes, remove the tubing from the fluid. Weigh and weigh again taking note of any physical changes to the tubing such as cracking, scarring, brittleness, or softening. These indicate the media damaged the tubing and would not be recommended. Work with quality control teams to determine a suitable change tolerance. Review system temperature and pressure next. Some formulations handle pressure better than others. Some are supported by infrastructure internal to the tubing such as braiding or mandrel wrapping. The braiding is inside the tube wall and mandrel wrapping is external, so they do not contact the process fluid. Wrapping and exterior stainless steel braiding typically offers a higher-pressure maximum than braided tubing. Alternatively, choosing a more rigid tubing formulation, such as PTFE or polypropylene, performs better than flexible tubing such as silicone at high system pressures. In addition to pressure, minimum and maximum temperatures for system and ambient to the tubing should be measured. Flexible tubing unsuitable for lower temperatures becomes brittle which may rupture or encourage spallation (the fragmentation of particulates from the inner surface of the tubing wall). At higher temperatures, the pressure capacity of the tubing decreases which can also lead to burst lines resulting in a potential safety hazard. Ideally, one will find a tubing formulation with a working temperature range where your process temperature is close to the mid-point. Purity of the formulation is also important to consider especially in downstream pharmaceutical and medical applications. Thankfully, there are many testing agencies that provide standards for purity and safety such as the Food & Drug Administration (FDA), the United States Pharmacopeia (USP), Registration, Evaluation, Authorization, and Restriction of Chemicals (REACH), and Restriction of Hazardous Substances (RoHS). Both REACH and RoHS are lists of chemicals determined to be harmful to human health or the environment by the European Union. These chemicals include lead, mercury and phthalate plasticizers such as DEHP. Both regulations come under review and receive updates on occasion. If biocompatibility is a concern, look for formulations certified as USP plastic class VI or simply USPVI. This classification outlines the parameters and guidelines for three main tests: systemic injection, intracutaneous injection, and implantation. The first two injecting a range of different biocompatible fluids such as saline and the latter simply being implanted into muscle. FDA 21 CFR is another that sets guidelines for the testing of materials intended for human end use. Most suppliers of high-grade tubing will supply a certificate of compliance (CoC) that will list the various regulatory approvals that the material meets. The certificate can even offer additional testing information as well. However, the latter testing information is usually gated content and is part of a more detailed validation packet. External considerations and partnering with your supplier External effects on the tubing are often overlooked compared to internal effects, but they are still pertinent. Silicone is moldable, so it is common that joints and flow intersection are molded together. However, not all flexible rubbers have this capability. Unfortunately, there is no rule of thumb whether one elastomer is weldable or not. The best solution is to partner with suppliers and inquire what tubing they can offer that is weldable. This is also true for sterilization technique. Silicone can withstand the most common sterilization techniques such as autoclave and gamma irradiation for some formulations. However, this is not the case for all alternative formulations. Overall, to ensure the correct alternative tubing is obtained, partner with a competent and resourceful tubing supplier on a solution. It is key to partner with a source that can review the complete fluid path and develop a plan to help ensure a continuous flow of necessary components to meet production time lines. By working through STAMP (size, temperature, application, media and pressure) of finding formulations and components that meet process needs, the supplier should also work on the necessary regulatory approval documentation, stocking, application support, and doing so from a global perspective (stocking locations around the globe). Let them help to confirm purity and compliance to regulations required by the process. How does one trust the recommendations or information from a supplier? The supplier should have comprehensive traceability and purity documentation of their formulations such as a certificate of compliance and validation guides—specific test data is also usually available upon request. Additionally, and perhaps most obviously, they should have quality trained support staff available to assist with any questions one may have regarding their products. Be flexible, like silicone rubber The siloxane supply constraint isn’t expected to be resolved until after 2019, so it would be wise for customers of silicone tubing to consider validating alternative formulations to assure continuity of product in their supply chain. This is not new information. Preparation and flexibility have always been good practices to follow whether there is a supply shortage or not. Making sure the process has some flexibility in its critical parameters mitigates unpredictability in supply forecasts and having some safety stock ensures no downtime if a shortage were to occur. Any supplier of quality tubing would be more than happy to collaborate on a process and help keep a business moving forward. |