Would you like to log in again?
Gastight® 1800 Series Syringes, Hamilton
Supplier: Hamilton
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
84875 84878 84881 84884 84887 84877 84880 84883 84886 84889 7659-01 7660-01 7661-01 7662-01 7663-01
60375-704EA 306.03 CAD InStock
60375-704 60375-726 60375-748 60375-760 60375-781 60375-806 60375-828 60375-840 60375-861 60375-883 10192-102 10192-104 10192-106 10192-108 10192-110
Gastight® 1800 Series Syringes, Hamilton
Syringes Syringes for Chromatography
The 1800 series is the reinforced plunger version of the 1700 series syringe. Attached to the flange of the syringe is a removable blue syringe holder that prevents heat transfer, dispense inaccuracies and plunger blow outs. It also provides support to small volume plungers that are prone to bending during injection.
  • Reinforced, protected plunger
  • Ideal for dispensing both liquids and gases in volumes from 1 up to 500 µL
  • Gas sterilisable
  • Stainless steel, borosilicate glass and PTFE fluid path
They have a precision machined PTFE plunger tip whichMore Product Information
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty
Print These Terms