Would you like to log in again?
Greiner Bio-One Immunology
Immunology
ELISA (Enzyme-Linked Immunosorbent Assay) is probably the most widely used biochemical method in laboratory analysis and diagnostics. Analytes such as peptides, proteins, antibodies and hormones can be detected selectively in low concentrations among a multitude of other substances and be quantified. Additionally, ELISAs are rapid, sensitive, cost-effective and can be performed in a high-throughput manner. ELISA is used in a variety of different assay types (e.g. direct ELISA, indirect ELISA, sandwich ELISA, competitive ELISA).