Would you like to log in again?
CE Compact Trolley
CE Compact Trolley
Compact size, full-size functionality! The CE Compact Trolley has been specifically designed for use in controlled environments where footprints are becoming smaller or where storage is a concern. The CE Trolley range has been specifically designed and developed for use in controlled environments where maximum control is needed. The trolleys are electro-polished stainless steel with airtight connections and smooth weldings. This ensures excellent biological control while still being easy to clean and fully autoclavable. The trolleys are extremely functional and versatile with superior ergonomics. All offer ample space for all utensils needed for effective cleaning and are fully customizable to meet your exact needs.
Learn More