Would you like to log in again?
Avantor OmniTop Sample Tubes® adjustable volume sampling system (AVSS)
Avantor OmniTop Sample Tubes® AVSS
Enhance the effectiveness of your sampling process with the Avantor OmniTop Sample Tubes® adjustable volume sampling system (AVSS). Designed to precisely collect small volume samples from your sterile process, the OmniTop Sample Tubes® AVSS helps eliminate volume loss in the connection lines, making it ideal for critical and high value product sampling in applications including:
  • Cell and gene therapy processes
  • Final fill operations
  • Glass and single-use bioreactor sampling
The OmniTop Sample Tubes® AVSS is single point or custom configurable into manifold designs. This novel solution is fully customizable to connect to your process and increase your product yields by reducing volume loss when compared to traditional methods of sampling. Choose from volumes ranging from 0.5 – 15.0mL to collect only what is needed for your analytical testing and meet the exact needs of your process.
Contact us today for more information and to learn how the OmniTop Sample Tubes® AVSS can make a difference in your operations.
Get more information