Would you like to log in again?
Solvents, LC-MS Grade, VWR Chemicals BDH
Solvents, LC-MS Grade
Our line of select LC-MS solvents is produced at the same state of the art ISO:9001 solvent manufacturing facility as our best-selling HPLC-grade line of solvents. These new solvents are specifically formulated for LC-MS, and feature high UV transmittance, low acidity and alkalinity, and low particulates to ensure minimal interference.
Product selection