Would you like to log in again?
Biotix Tubes
High Quality tubes for your everyday labwork
PCR Tubes
Biotix PCR tubes are manufactured under stringent QC protocols with medical-grade virgin polypropylene. These tubes consist of a thin and uniform wall thickness that provides equal thermal transfer though the tube for optimal amplification. PCR tubes are available in single or 8 strip format, caps sold separately.
Purchase our PCR tubes here
Microcentrifuge Tubes
Biotix microcentrifuge tubes are manufactured with medical grade virgin polypropylene for superior clarity. These snapcap tubes are leak resistant, boil proof, and able to withstand 20000 RCF. Frosted exterior walls and cap make for easy marking and sample identification. Available in low binding and multiple sizes.
Click here to view out MCT options