Would you like to log in again?
Chemical Resistance/Flame Resistance
DuPont™ ProShield 6 SFR
  • Cost-efficient disposable overgarment designed to help protect and preserve primary flame resistant garments
  • Intended to be worn over primary flame resistant garments to provide a barrier against non-hazardous particles and aerosols
  • Designed to not contribute to additional burn injury in the event of an industrial fire or electric arc event
  • Will not ignite and continue to burn after the flame source is removed
  • Excellent choice as a cost efficient cover to protect reusable FR garments
  • Applications include: Steel Mills, Coal Mines, Maintenance Work, Refineries, Laboratories, Utilities
Learn more
DuPont™ Tychem® 2000 SFR
  • Provides chemical and secondary flame protection in a lightweight garment
  • Intended to be worn over primary flame resistant garments
  • In the event that a flash fire occurs, they won't ignite and won't contribute to additional burn injury if appropriate flame-resistant personal protective equipment, such as DuPont™ Nomex®, is worn beneath
  • Provides an effective barrier against a range of inorganic acids and bases as well as industrial cleaning chemicals and particles
  • Ideal option for keeping workers safe from chemicals, while not compromising the flame protection of fire resistance clothing worn beneath them
  • Applications include: refineries, petrochemical plants, laboratories, hazardous maintenance operations
Learn more