Would you like to log in again?
Cut resistant gloves
Cut resistant gloves
Wells Lamont Industrial specializes in cut resistant gloves with many different products.  From fabric to palm dipped to metal mesh gloves we cover many different applications and levels of cut resistance.
Thin glove liner with great dexterity and ANSI A4 cut resistance
The M121 glove liner offers ANSI 4 cut resistance and is highly reusable with great dexterity. Made with antimicrobial fibers.
Metal mesh glove
This glove offers superior cut resistance and features a patented self tensioning cuff that is easier to put on and more hygenic.
Palm dipped glove
The Y9265 has a polyurethane palm for grip and offers ANSI A2 cut resistance.
Sleeve with ANSI A7 cut resistance
The Armguard II Medium Duty is a 20" high level cut resistant sleeve that is made in the USA.
Fully dipped glove with ANSI A3 cut resistance
The Y9290 offers a solid grip and ANSI A3 cut resistance to protect your hands from liquids while offering superior protection.
Silver Talon- ANSI A6 cut resistance
The Silver Talon is approved for food contact, low lint, and is a great glove to protect your hands from extreme hazards. Made in the USA.