Would you like to log in again?
Performance level 1: Everyday analysis, Chromatography approved
Performance level 1: Everyday analysis, Chromatography approved
Choose the right vial for chromatographic analyses. Thermo Scientific SureSTART Performance level 1 products are designed for everyday chromatography analysis. They provide an affordable choice for use with all GC and HPLC/UHPLC instrument types. If you are verifying compound presence and quantification using detection techniques such as UV/Vis, PDA,  RI, ELSD, FLD, FID, NPD, ECD, TCD, or FPD, our Performance level 1 products are the perfect solution for you– select the products with the one wave.
Within Performance level 1 you will find a selection of vials (glass, plastic, <2 mL, 2 mL and headspace). You can also find a great selection of standard and bonded caps to support robotic autosamplers within this performance level. Need stoppers? These can also be found within this performance level.
  • Packaged in material that has been tested and selected for cleanliness
  • All products within this performance level are suitable for use with GC and HPLC/UHPLC
Specification certified
  • All Thermo Scientific SureSTART vials are dimensionally verified to ensure autosampler compatibility
  • Automated optical and gauge inspection to ensure critical dimensions and tolerances
  • Vials meet all relevant requirements of US, EU, JPN Pharmacopeia
  • Uniform and defect-free
Lot traceable
  • 100% traceable through the manufacturing process
  • Certificate of Conformance available by lot number upon request
Performance level 1 products include: 
SureSTART Vials/ Closures  
Glass: Amber/Clear
Screw 2mL /9mm Screw Caps
Crimp 2mL /11mm Crimp Caps (Magnetic Steel and Aluminum)
Headspace Crimp 20 mL /20mmCaps
Plastic: Polypropylene (PP) Microvials for <2mL
Screw Top Plastic: Polypropylene (PP) Microvials for <2mL
Snap Top Plastic: Polypropylene (PP) Microvials 11mm for <2mL
Stoppers
20 mm stoppers
Crimpers, Decrimpers and Decapping
Crimpers, Decrimpers and Decapping Pliers 
Accessories for Programmable Electronic High-Power Crimp Stations
Electronic crimpers and decrimping
SureSTART Well Plates, Mats, Sealing Tapes, Handheld Applicators
WebSeal well plates:  Plastic, non-coated, non-sterile, chromatography tested
WebSeal Mats and Sealing Tapes, Non sterile with Handheld Applicators