Would you like to log in again?
Generating CRISPR cell models, an overview, challenges and latest technologies
CRISPR/Cas9 is a revolutionary technology to irreversibly and accurately modify the genome. To successfully perform genome editing it is essential to understand which techniques are available to deliver CRISPR/Cas9 components into cells. As transfection experts, Polyplus-transfection gives an overview of how CRISPR/Cas9 achieves precise genome editing, delves into key challenges and considerations that come with delivery of CRISPR/Cas9 components into cells, and presents latest technologies to address these challenges. The webinar will finish with a short Q&A session, with the opportunity for follow-up discussions.
Polyplus-transfection offers a large range of transfection reagents that cover available CRISPR approaches:
  • Transfection of plasmid DNA encoding Cas9, gRNA and gene of interest (jetPRIME and jetOPTIMUS)
  • Transfection of gRNA and mRNA encoding Cas9 (jetMESSENGER) 
  • Delivery of RNP, meaning the Cas9 protein complexed with the gRNA (jetCRISPR and SpCas9 nuclease)
More Info
Thursday,
29th November 2018,
From 15:00 to 16:00 BST
From 16:00 to 17:00 CET
  • Email alerts before the webinar
  • Free registration
  • Chat area to ask questions
  • The webinar can be replayed for 1 year
Time Zone: BST
(Dublin, Edinburgh)
Language:
English  
Register