Would you like to log in again?
TRIO.BAS - The most complete line of impact microbial air sampling instruments available
TRIO.BAS Microbial Air Samplers
TRIO.BAS Microbial Air Samplers are available in Petri, Contact plate, or Compact Dry formats, 100-200L per minute models, and a variety of ergonomic, lightweight platforms that make active impact air monitoring a breeze. Battery life of 15 hours on a single charge and no plugs or external connections reduce the risk of contamination.  Always expanding, the TRIO.BAS line offers the most utility of any air sampler with the addition of the new AIRBIO DUO, RABS, and Multiflex models.
TRIO.BAS, the future of air sampling here today!