Would you like to log in again?
XPert® Balance Systems & Enclosures
 XPert® Balance Systems & Enclosures
Labconco's™ industry leading XPert Balance Systems and Enclosures provide user protection by keeping powders and particulates contained during manipulations such as weighing. The System features a built-in blower and exhaust HEPA filter and takes up less space on the bench. Vibration isolation allows for steady weighing with a microbalance. Enclosure models are flexible and may be hooked up to a portable exhauster or filtered house exhaust.
Now through the REDISHIP program, Labconco XPert Systems and Enclosures are available within 48 hours.
Certifications: Independently validated by SafeBridge Consultants. Tested to Modified ASHRAE 110. ETL listed. Contact Avantor to see how you can maximize protection from hazardous powders while minimizing space requirements.