Would you like to log in again?
Leica Electron Microscopy Sample Preparation
Leica Electron Microscopy Sample Preparation
Ensure the ultrastructures of your tissue samples can be precisely compared every time by preparing them with the EM TP tissue processor.
With the Electron Microscopy Tissue Processer (EM TP) you can be confident that tissue differences between samples observed with your light microscope (LM) or electron microscope (EM) are not caused by inconsistent manual preparation. The EM TP limits manual handling by at least 75% due to programmable, automatic processing of multiple samples.