Would you like to log in again?
Masterflex® single-use components
Choose reliable components adhering to strict sterilization protocols
With Masteflex single-use, you will no longer have to validate the cleaning and sterilizing of your bioprocess assemblies. The purchase of a preassembled, sterilized and validated system shifts the burden of validation to the supplier. That is why it is important to be sure your supplier uses only the most reliable brand-name products and adheres to strict, well-established protocols in the sterilization process.
Masterflex single-use assemblies are manufactured in a Class 10000 cleanroom. The assembly facility is ISO 13485 certified and are cGMP compliant. We offer gamma irradiation sterilization with a 10–6 SAL (Sterility Assurance Level), a one millionth probability of microbial survival, and full lot traceability that identifies the production site, manufacturing date and origin of the components. We also provide and maintain all required documentation, including complete validation documents and certificates of conformance and sterility.
We build custom assemblies with reliable products
Fittings
Tubing
Sensors
Common accessories
It’s time to make your process more efficient
  1. What you need
    Choose from thousands of components. We custom configure, validate and package according to your specifications.
  2. When you need it
    Stop cleaning and revalidating your system. We manage your inventory and send assemblies according to your schedule.
  3. Call for a quote today
    Masterflex single-use experts provide a quote with full schematic, followed by an actual sample for your approval.