Fire Assay Flux
|
As a leading manufacturer of high-purity fire assay flux, our experience has earned us an exceptional reputation for offering fire assay flux of the highest quality to the global assay industry. We utilize strict quality control standards that guarantee gold values of less than 1.5 ppb in litharge component of the flux. Assay flux with low silver levels (Gold and Silver Free) is also available. Flux comes in either a fine powder or granular format, both of which vary in color between light yellow and orange. We offer customer-specific formulations that will satisfy the most demanding specifications while adhering to the strictest confidentiality for proprietary customer knowledge. A Certificate of Analysis is available for each batch of flux manufactured. Packaging is available in 11.5 kg, 20kg, 25kg, or 40kg UN-approved plastic pails or 500kg UN-approved maxi bags. |
VWR offers a wide assortment of products and brands in this category. If you cannot find the product or brand that you are looking for in this listing, please use the general search field in the top-right corner of the screen to search the entire VWR portfolio.
You can also contact your local VWR/Anachemia office or sales representative.
Flux Ingredients
-
Borax, Anhydrous
- Flour 10KG Bag
-
Fluorspar
-
Inquarts, Herman
-
Lead Foil
-
Litharge, Lead (II) oxide, granular Gold and Silver Free
-
Litharge, Lead (II) oxide, powder Gold and Silver Free
-
Potassium Nitrate
-
Potassium Nitrate, crystals, Reagent, ACS
-
Silica Flour, 200 Mesh, Lab Grade
-
Silica Sand, Granulated, 30 Mesh, Lab Grade
-
Silver Foil, 99.99%, Refining Systems
-
Silver Nitrate, crystals, Reagent, ACS
-
Silver Sulfate, powder, Reagent, ACS
-
Soda Ash 58%, Light, Granulated, Lab Grade
-
Soda Ash, Dense, Granulated, Lab Grade
- Test Lead, Granulated