Would you like to log in again?
Azure Cielo™ 3 and Cielo 6
Free 7-Day qPCR Trial
The Azure Biosystems™ Cielo™ 3 and 6 real-time PCR Systems are designed to offer smart protocols, high quality data, robust durability and an intuitive user experience.
Our novel fiber optic detection system allows for enhanced sensitivity and speed. 3 or 6 standard channels let you analyze a variety of commercially available dyes, already in your workflow. Onboard software operated by a 10.2″ touchscreen minimizes footprint and saves you bench space.
Easily connect to your Cielo via Wi-Fi, ethernet, USB or LAN. Get notified after each completed run and even remotely access protocols and files. Contact us for to demo the Cielo for 7 days- no risk, no cost to you!