Would you like to log in again?
Chemical Label Information
Ward's Chemical Labels
Every Ward's chemical label meets the latest OSHA/WHMIS labeling requirements in a clear and concise format and include the following information:
  • Product identifier - chemical name, grade, and properties
  • Supplier identification - name, address, and contact information
  • Pictograms - indicates specific information about chemical hazard(s)
  • Signal word - indicates severity of hazard(s)
  • Hazard statements - describes nature of hazard
  • Precautionary statements - recommended measures to prevent adverse effects
Example of a Ward's Chemical Label:
For help reading your Ward's chemical label, or assistance and materials for updating your existing chemical labels to match the latest requirements, contact the Ward's Science Plus Us Chemistry team at sciencehelp@vwr.com.