Would you like to log in again?
Automation Tips for Liquid Handling Systems
Automation Consumables
With hundreds of Thermo Scientific™ automation tips designed to fit over 50 workstations and pipetting head variations, we're certain you will find tips that increase efficiency through accurate and precise performance.
These tips undergo a rigorous quality control program to ensure consistency resulting in superior tip straightness and a low coefficient of variation ranging from 1% to 3%. With stringent manufacturing specifications, our tips adhere to tolerances 400% tighter than standard pipette tips.
Thermo Scientific Automation Tips for Tecan Automated Liquid Handling Platforms
  • Conductive option provides increased pipetting performance
  • Tips are validated on the actual workstations for which they are created
  • 20 μl tips increase precision of low volume pipetting through minimization of dead air space
Thermo Scientific Automation Tips for PerkinElmer/Packard Robotic Automated Liquid Handling Platforms
  • Conductive option provides increased pipetting performance
  • Increased precision for your toughest multi-dispensing assays
  • Tips are validated on the actual workstations for which they are created
Thermo Scientific Automation Tips for Agilent Automated Liquid Handling Platforms
  • Low percentage coefficient of variation
  • Available in nonsterile, sterile or sterile filtered
  • Validated on the actual workstation for which the tip is created
Thermo Scientific Automation Tips for Eppendorf Automated Liquid Handling Platforms
  • Low percentage coefficient of variation
  • Available in nonsterile, sterile or sterile filtered
  • Validated on the actual workstation for which the tip is created
Thermo Scientific Automation Tips for other Automated Liquid Handling Platforms
Tips for:
  • Qiagen Universal Systems
  • Molecular Devices Systems
  • Caliper/Zymark Liquid Handling Workstations
  • BioTek Pipetting Systems
  • Thermo Scientific Automated Liquid Handling Platforms