J.T.Baker® Viral Inactivation SolutionViral Inactivation with high purity and qualityMaximize downstream efficiency and minimize riskThe new readily biodegradable J.T.Baker® Viral Inactivation Solution from Avantor® is a highly effective detergent that efficiently inactivates or destroys residual viruses prior to the Protein A capture step. Compatible across a range of molecules, J.T.Baker® Viral Inactivation Solution minimizes contamination risk by reducing the viral burden on subsequent process steps without impacting protein integrity. Superior efficiency & performanceJ.T.Baker® Viral Inactivation Solution provides comparable viral clearance to Triton X-100 while eliminating processing steps, increasing efficiency in your downstream process.
High purity reduces downstream riskImplement detergent-based viral inactivation prior to the protein capture step to decrease viral burden downstream and overcome environmental concerns.
Ease of use offers resource savingsAchieve cost, time and labor savings with minimized processing challenges associated with foaming and viscosity, less required detergent volume and no viral inactivation buffer prep.
J.T.Baker® Viral Inactivation Solution![]() Seeking a sustainable solution for viral inactivation? Technical NotesArticlesWebinars |