Would you like to log in again?
NEW VWR® Hotplates and Stirrers
NEW VWR® Hotplates and Stirrers
The VWR Advanced and Professional Hotplate-Stirrers have been redesigned with safety as a priority. Industry leading heat monitoring and shutoff features promote a safe laboratory environment.  World-Class temperature capabilities coupled with powerful stirring make these models the right choice. All heating models feature a connection for a temperature probe and have calibration functions. Assembled in the USA, including a best-in-class 5 year warranty. Hotplate-Stirrers are available in 7x7", 10x10", and 135mm diameter round top.
Select Professional models when enhanced features such as a timer, temperature limiting function, and monitoring the actual versus set temprature are needed and includes a PT1000 RTD temperature probe. 
Choose the right model for your application!