Would you like to log in again?
CE Duo® CleanTech Duo® Sponge
CE Duo® CleanTech Duo® Sponge
Microfiber cleaning and disinfecting for hard to reach areas! The CE Duo™ CleanTech™ Sponge allows you to disinfect while cleaning hard-to-reach areas in cleanrooms and controlled environments. The patented microfiber composition removes contamination from surfaces and holds it within the sponge. The microfiber laminated foam is ideal for cleaning and disinfecting all types of surfaces and hard-to-reach areas, such as behind equipment, cabinets and piping, as well as hoods, isolators, door handles, transition surfaces and all other surfaces where a mop cannot reach. The sponge can be used with either the bucket or pre-prepared method. The double sided, hand-held design results in a higher degree of user control.
Learn More