Would you like to log in again?
Greiner Bio-One Liquid Handling
Liquid Handling
Pipette Tips are a key component in day-to-day life science research and need to be of a high quality to ensure confidence and consistency in pipetting results. Our tips are manufactured from high-grade polypropylene to give the optimum in performance and fit.
Additionally, polypropylene has a high breaking strength, is dimensionally stable, heat-resistant up to approx. +140 °C, i.e. is autoclavable, and cold-resistant down to -190 °C. All colored Greiner Bio-One pipette tips are manufactured exclusively with heavy metal free dyes.
Pipette tips from Greiner Bio-One are therefore free of heavy metals.
The Sapphire product family comprises standard, filter and low retention tips manufactured from virgin materials. All tips are transparent, graduated, and allow precise and comfortable pipetting with maximal recovery.