Would you like to log in again?
Greiner Bio-One CELLSTAR® Tissue Culture
Cell & Tissue Culture
Greiner Bio-One CELLSTAR® products offer a wide spectrum of cell culture flasks, dishes and plates. Their functional design, particular material properties and special coatings enable these vessels to be used not only in standard cell culture but also in cancer research, viral diagnostics, genetic engineering and vaccine production.
In response to the increasing automation of cell culture procedures, Greiner Bio-One developed the AutoFlask™, a cell culture flask for automated systems. With the ThinCert™Plate and appropriate cell culture inserts, Greiner Bio-One has also created an innovative platform for compatibility tests using skin models in the chemical and cosmetic industries.
The Cell Culture product sector was recently expanded with the innovative Advanced TC™ polymer modification. These cell culture products have a special surface that enables more effective cultivation of fastidious cells and facilitates complex testing systems
For high-resolution microscopy and live cell analysis, Greiner Bio-One offers the new CELLview™ dish, a polystyrene cell culture dish with an embedded autofluorescence-free glass bottom. The unique subdivided version of the dish is ideally suited for multiplex analyses, for example, and is also available with the Advanced TC™ surface.
Product Selection