Would you like to log in again?
Best DNA transfection reagent - jetOPTIMUS®
Best DNA transfection reagent - jetOPTIMUS®
jetOPTIMUS® is a powerful transfection reagent that improves cellular uptake and endosomal escape of DNA in adherent cells (even in hard-to-transfect cells) resulting in higher transfection efficiency.
  • Highly efficient: reach maximal gene expression in hard-to-transfect cells
  • Cost-effective: use minimum reagent volume and DNA quantity
  • Biologically relevant: keep an excellent cell viability & morphology
  • Time-saving: transfect with an optimized ready-to-use protocol
Request your free sample today
Description
jetOPTIMUS® is an innovative cationic nanotechnology developed to improve DNA transfection efficiency in difficult-to-transfect cells used as in vitro cell culture models. Tested on various primary cells and cell lines, jetOPTIMUS® proved its superiority by reaching higher transfection efficiencies and gene expression than main competitors.
jetOPTIMUS® outperforms its main competitor. Transfection efficiency was assessed by FACS analysis in various cell lines 24 h after transfection with a plasmid coding for GFP protein in 24.well plates. Conditions were set up according to the manufacturer's recommendations both for Lipofectamine ® 3000 and jetOPTIMUS ®.
jetOPTIMUS® has been developed to increase transfection efficiency into a wide range of cell types, while keeping the amount of DNA as low as possible, and reducing the volume of reagent required compared to main competitors. Consequently, scientists can increase the number of transfections and reduce the cost per reaction by up to seven-fold.
Lower volume of jetOPTIMUS® is required compared to Lipofectamine® 3000 in 24-well plate. Consequently, scientists can increase the number of transfections and reduce the cost per reaction by up to seven-fold.
jetOPTIMUS® transfection reagent is also very gentle on cells, allowing generation of biologically relevant data from gene expression studies.
Cells remains healthy and keep a good morphology 24h after transfection with jetOPTIMUS®. Phase contrast microscopy of both MDCK and VERO cells 24 h after transfections performed according to the manufacturer's recommendations for each reagent.
jetOPTIMUS® is extermely easy to use and is provided with its own buffer.
Testimonials
“In comparison to another transfection reagent, jetOPTIMUS was mind-blowingly better for the difficult to transfect 3T3-NR6 cells!“
– Danai L., Katholieke Universiteit Leuven, BE
“Very easy to use, sensitive cell lines support it very well, good efficiency and reproducibility, I would recommend it.”
– Huguette D., UZH, CH
“jetOPTIMUS is a great product. It worked better than competitors to deliver large plasmid constructs into hard-to-transfect cells.”
– Mona S., University, DE