Would you like to log in again?
Battery Materials Production
Battery Materials Production
Avantor sets advanced battery science in motion to create a better world
Current Li-ion batteries consist of largely four main components: cathode, anode, electrolyte, and separator.
  • The cathode is a thin aluminum foil coated with a compound made up of active agent, solvent, super conductive additive and binder. Cathode active materials are typically composed of lithium and metal.
  • The anode is typically a thin copper foil coated with a compound made up of active agent, solvent, super conductive additive and binder. Anode active material is graphite (synthetically-produced or mined).
    The electrolyte is composed of salts, solvents and additives.
  • Typical separators are currently made of synthetic resin such as polyethylene (PE) and polypropylene (PP)
Avantor will support you by offering critical raw materials, chemicals, equipment, instruments as well as a broad range of products for R&D, QA/QC, Testing, controlled environment, personal protective equipment, safety and facility management
Contact your local sales representative for more information or fill in our contact form
Controlled Environment
Avantor offers a broad range of cleanroom products to protect the product and process.
Protective Equipment & Safety
Avantor offers a broad range of Personal Protective Equipment (PPE) to protect the operator when researching, manufacturing or recycling.
Facility Management
Avantor offers a broad range of products to keep the facility clean, safe and ordered.
Chemicals & Raw Materials
Avantor offers a broad range of chemicals for R&D and production of battery active materials:
Equipment & Instruments
Avantor distributes a broad range of process equipment and instrumentation from top manufacturers.
R&D, QA/QC. Testing
Avantor carries a broad range of products to meet the specialized needs of R&D, QA/QC & Testing laboratories.
Featured Products – Chemicals & Raw Materials
LiPF6
The most common electrolyte in lithium batteries is a lithium salt solution such as lithium hexafluorophosphate (LiPF6)
Featured Products – Personal Protective Equipment & Safety
Protective apparel
Discover our extensive range of workwear, disposable coveralls…
Fall Harnesses
Personal fall arrest systems
Gloves
Refine here the search of the right glove for your application
Gas Detectors
Multi-gas detectors, oxygen detectors.
Featured Products – Facility Management
Chemical Storage
Our chemical storage solutions provide safe way to store corrosive or flameable materials, protecting personnel and facilities...
Spill Control
Our wide range of stationary systems and handheld wiping units can help you keep your laboratory space clean of spilled chemical solution or run-off. Look at the product Avantor offers for general-purpose spill control or specific applications.