Would you like to log in again?
Titration and Karl Fischer determination
Titration and Karl Fischer determination
Titration and Water determination are really common in all R&D and QC Labs. To perform in these 2 methods, you need a large range of accurate products.
Volumetric solutions
Titration is a technique that enables you to determine the precise concentration of everything. You will find below a wide range of accurate solutions in different formats (Bottles and bag-in -box).
Karl Fischer reagents
Wide range of reagents for the determination of water by Karl Fischer titration. These include reagents for one- or two-component systems; combined coulometric reagents; solvents for oils and fats; reagents and solvents for determination of water in...
Karl Fischer Standards
Standards for Karl Fischer titration can be used for monitoring and qualification of your Karl Fischer instruments, for checking and evaluating measuring results, and for performing titer determination of volumetric Karl Fischer reagents.