Would you like to log in again?
Downstream purification for cell therapy
Comparing downstream purification approaches between cell therapies and other biologics is quite different.
Methods required to obtain the final cell dose with the required quality parameters include: 
  • Centrifugation for removal of debris, washing, and concentration of the cell dose, 
  • Alternatives to centrifugation, such as counter-flow centrifugation, and 
  • Depth and ultrafiltration using TFF options.
Explore solutions for downstream purification of cell therapy
Explore product choices for downstream purification of cell therapy
Ready to Maximize Production Efficiency?
Our Bioprocessing Solutions team can help you streamline your recombinant protein processes!
Get Started