Would you like to log in again?
BRAND HandyStep Repeating Pipettes & PD-Tips II Syringe Tips
Repetitive Pipettors & Tips
The NEW BRAND HandyStep Touch Motorized Repeating Pipette features an easy-to-use touchscreen interface for intuitive operation. HandyStep Touch features dispensing, auto-dispensing, and single volume pipetting modes, with the ability to save up to 10 favorite functions. The HandyStep Touch S adds multi-aspiration, sequential dispensing, and titration modes for even more versatility. The HandyStep Touch and Touch S automatically recognizies BRAND PD-Tip II precision dispenser tips, and also features an open design to accommodate all standard type repeating pipette tips.
NEW BRAND PD-Tip II syringe tips are made from high-quality virgin plastics for us in all standard repeating pipettes in most applications. Their precision seal ensures smooth operation with increased accuracy and precision, while the extended piston minimizes dead volume and maximizes fluid utilization. PD-Tip II tips are encoded for auto-recognition in the BRAND HandyStep Touch, along with other major electronic repeating pipettes.