Would you like to log in again?
BRAND Pipet Controllers
Pipet Controllers
BRAND accu-jet pro and macro pipette controllers provide fine control over volumetric and serological pipettes from 0.1 mL to 200 mL in size, with superior ergonomics. PTFE ball valves allow smooth control from dropwise to full flow. A speed limiter on the accu-jet pro facilitates filling of small volume pipettes. PTFE membrane filters (0.2 µm for accu-jet pro, 3 µm for macro) help protect samples and controller components.