Would you like to log in again?
Captair Filtering Chemical Storage Cabinets
Captair Filtering Chemical Storage Cabinets
Erlab’s CaptairStore Filtering Chemical Storage Cabinets will store your active use bottles within a filtered chemical storage cabinet, operating 24 hours 7 days a week. The filter within will ensure no buildup of chemical vapors or odors within the cabinet. No HVAC connection to the outside will be required. Each cabinet has Smart Technology™ - an exclusive set of tools that consists of Smart-Light Communication™, chemical sensors (*optional), real-time status, and the eGuard app.
  • Smart Technology gives an easily identifiable method of communication regarding containment, performance and filter efficiency via a soft, LED band of light called Smart-Light
  • Smart-Light is a light signature across the front panel which casts a stable LED glow ensuring proper operation (LED Lighting > 650 Lux)
  • Corrosion resistant electro-galvanized steel coated with anti-acid polymer Clear, chemical resistant acrylic doors for easy viewing